MilliporeSigma
  • Nanoparticles made from novel starch derivatives for transdermal drug delivery
Nanoparticles made from novel starch derivatives for transdermal drug delivery
Journal of Controlled Release : Official Journal of the Controlled Release Society (2010)
Santander Ortega MJ, et al.