MilliporeSigma
  • A TgCRND8 mouse model of Alzheimer?s disease exhibits sexual dimorphisms in behavioral indices of cognitive reserve
A TgCRND8 mouse model of Alzheimer?s disease exhibits sexual dimorphisms in behavioral indices of cognitive reserve
Journal of Alzheimer'S Disease (2016)
Granger MW, et al.