MilliporeSigma
  • Primary structure of a human mitochondrial protein homologous to the bacterial and plant chaperonins and to the 65-kilodalton mycobacterial antigen.
Primary structure of a human mitochondrial protein homologous to the bacterial and plant chaperonins and to the 65-kilodalton mycobacterial antigen.
Molecular and cellular biology (1989-05-01)
S Jindal, A K Dudani, B Singh, C B Harley, R S Gupta
ABSTRACT
The complete cDNA for a human mitochondrial protein designated P1, which was previously identified as a microtubule-related protein, has been cloned and sequenced. The deduced amino acid sequence of P1 shows strong homology (40 to 50% identical residues and an additional 20% conservative replacements) to the 65-kilodalton major antigen of mycobacteria, to the GroEL protein of Escherichia coli, and to the ribulose 1,5-bisphosphate carboxylase-oxygenase (rubisco) subunit binding protein of plant chloroplasts. Similar to the case with the latter two proteins, which have been shown to act as chaperonins in the posttranslational assembly of oligomeric protein structures, it is suggested that P1 may play a similar role in mammalian cells. The observed high degree of homology between human P1 and mycobacterial antigen also suggests the possible involvement of this protein in certain autoimmune diseases.