MilliporeSigma
  • Genetic Variations of the COL4A1 Gene and Intracerebral Hemorrhage Risk: A Case-Control Study in a Chinese Han Population
Genetic Variations of the COL4A1 Gene and Intracerebral Hemorrhage Risk: A Case-Control Study in a Chinese Han Population
World Neurosurgery (2018)
Lin S, et al.