MilliporeSigma
  • Phenotype variability in a large Spanish family with Alport syndrome associated with novel mutations in COL4A3 gene
Phenotype variability in a large Spanish family with Alport syndrome associated with novel mutations in COL4A3 gene
BMC Nephrology (2017)
Cervera AC, et al.