MilliporeSigma
  • Transcriptome characteristics and X-chromosome inactivation status in cultured rat pluripotent stem cells
Transcriptome characteristics and X-chromosome inactivation status in cultured rat pluripotent stem cells
Stem Cells and Development (2015)
Vaskova EA, et al.