MilliporeSigma
  • Morphology of the myoepithelial cell: immunohistochemical characterization from resting to motile phase
Morphology of the myoepithelial cell: immunohistochemical characterization from resting to motile phase
TheScientificWorldJournal (2012)
Beha G, et al.