MilliporeSigma
  • The Toll-Like Receptor/MyD88/XBP1 Signaling Axis Mediates Skeletal Muscle Wasting during Cancer Cachexia.
The Toll-Like Receptor/MyD88/XBP1 Signaling Axis Mediates Skeletal Muscle Wasting during Cancer Cachexia.
Molecular and cellular biology (2019-05-30)
Kyle R Bohnert, Praneeth Goli, Anirban Roy, Aditya K Sharma, Guangyan Xiong, Yann S Gallot, Ashok Kumar
ABSTRACT
Skeletal muscle wasting causes both morbidity and mortality of cancer patients. Accumulating evidence suggests that the markers of endoplasmic reticulum (ER) stress and unfolded protein response (UPR) pathways are increased in skeletal muscle under multiple catabolic conditions, including cancer. However, the signaling mechanisms and the role of individual arms of the UPR in the regulation of skeletal muscle mass remain largely unknown. In the present study, we demonstrated that gene expression of Toll-like receptors (TLRs) and myeloid differentiation primary response gene 88 (MyD88) was increased in skeletal muscle in a Lewis lung carcinoma (LLC) model of cancer cachexia. Targeted ablation of MyD88 inhibits the loss of skeletal muscle mass and strength in LLC tumor-bearing mice. Inhibition of MyD88 attenuates the LLC-induced activation of the UPR in skeletal muscle of mice. Moreover, muscle-specific deletion of X-box binding protein 1 (XBP1), a major downstream target of IRE1α arm of the UPR, ameliorates muscle wasting in LLC tumor-bearing mice. Our results also demonstrate that overexpression of an active form of XBP1 caused atrophy in cultured myotubes. In contrast, knockdown of XBP1 inhibits myotube atrophy in response to LLC or C26 adenocarcinoma cell conditioned medium. Collectively, our results demonstrate that TLR/MyD88-mediated activation of XBP1 causes skeletal muscle wasting in LLC tumor-bearing mice.