MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Rice and wheat straw ashes: Characterization and modeling of pretilachlor sorption kinetics and adsorption isotherm.
Rice and wheat straw ashes: Characterization and modeling of pretilachlor sorption kinetics and adsorption isotherm.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2019-01-15)
Anup Kumar, Abhishek Mandal, Neera Singh
ABSTRACT
The rice straw ash (RSA) and wheat straw ash (WSA) were explored as low cost adsorbent for pretilachlor removal from water. The ashes were characterized and sorption behavior of pretilachlor was evaluated. Kinetics study suggested that the modified Elovich model best explained the pretilachlor sorption on both the ashes. The adsorption data were analyzed using 2-, 3- and 4-parameter models and nine error functions were used to compute the best fit isotherm by nonlinear regression analysis. The pretilachlor was more sorbed onto the RSA (22.0-92.2%) than the WSA (11.4-61.4%) and percent adsorption decreased with increase in the herbicide concentration in solution. Isotherm model optimization analysis suggested that the Freundlich and the Temkin isotherms were the best models to predict the pretilachlor adsorption onto the RSA and the WSA. The error analysis suggested that the reciprocal of the observed squared (ROS) and the reciprocal of the predicted squared (RPS) error functions provided the best determination of the adsorption constants for the Freundlich and the Temkin isotherms, respectively. The RSA, which exhibited higher pretilachlor sorption potential, can be utilized as low cost adsorbent for pesticide removal from contaminated water.