MilliporeSigma
  • Ozone-Based Advanced Oxidation Processes for Primidone Removal in Water using Simulated Solar Radiation and TiO2 or WO3 as Photocatalyst.
Ozone-Based Advanced Oxidation Processes for Primidone Removal in Water using Simulated Solar Radiation and TiO2 or WO3 as Photocatalyst.
Molecules (Basel, Switzerland) (2019-05-07)
Manuel A Figueredo, Eva M Rodríguez, Manuel Checa, Fernando J Beltran
ABSTRACT
In this work, primidone, a high persistent pharmacological drug typically found in urban wastewaters, was degraded by different ozone combined AOPs using TiO2 P25 and commercial WO3 as photocatalyst. The comparison of processes, kinetics, nature of transformation products, and ecotoxicity of treated water samples, as well as the influence of the water matrix (ultrapure water or a secondary effluent), is presented and discussed. In presence of ozone, primidone is rapidly eliminated, with hydroxyl radicals being the main species involved. TiO2 was the most active catalyst regardless of the water matrix and the type of solar (global or visible) radiation applied. The synergy between ozone and photocatalysis (photocatalytic ozonation) for TOC removal was more evident at low O3 doses. In spite of having a lower band gap than TiO2 P25, WO3 did not bring any beneficial effects compared to TiO2 P25 regarding PRM and TOC removal. Based on the transformation products identified during ozonation and photocatalytic ozonation of primidone (hydroxyprimidone, phenyl-ethyl-malonamide, and 5-ethyldihydropirimidine-4,6(1H,5H)-dione), a degradation pathway is proposed. The application of the different processes resulted in an environmentally safe effluent for Daphnia magna.