MilliporeSigma
  • Distinct Expression and Clinical Significance of Zinc Finger AN-1-Type Containing 4 in Oral Squamous Cell Carcinomas.
Distinct Expression and Clinical Significance of Zinc Finger AN-1-Type Containing 4 in Oral Squamous Cell Carcinomas.
Journal of clinical medicine (2018-12-14)
Julián Suárez-Canto, Faustino Julián Suárez-Sánchez, Francisco Domínguez-Iglesias, Gonzalo Hernández-Vallejo, Juana M García-Pedrero, Juan C de Vicente
ABSTRACT
Zinc finger AN1-type containing 4 (ZFAND4) has emerged as a promising prognostic marker and predictor of metastasis for patients with oral squamous cell carcinoma (OSCC). However, further validation is fundamental before clinical implementation. Hence, this study evaluated the expression pattern of ZFAND4 protein expression by immunohistochemistry using an independent cohort of 125 patients with OSCC, and correlations with the clinicopathologic parameters and disease outcome. Remarkably, ZFAND4 expression, while negligible in normal epithelium, exhibited two distinct expression patterns in tumors that did not overlap. A gross granular staining was characteristic of the undifferentiated cells at the invasive front of tumors, whereas the most differentiated cells located at the center of the tumor nests showed diffuse non-granular staining. ZFAND4 staining was higher in undifferentiated than in differentiated areas of tumors. High ZFAND4 expression in differentiated cells was significantly associated to well-differentiated (p = 0.04) and non-recurrent tumors (p = 0.04), whereas ZFAND4 expression in undifferentiated cells correlated with tumor location (p = 0.005). No correlations between the ZFAND4 expression and patient survival were found. These data question the clinical relevance of ZFAND4 expression as a prognostic biomarker in OSCC, and also reveal distinct ZFAND4 expression patterns depending on the differentiation areas of tumors that should be evaluated separately.