MilliporeSigma
  • Possible biased analgesic of hydromorphone through the G protein-over β-arrestin-mediated pathway: cAMP, CellKey™, and receptor internalization analyses.
Possible biased analgesic of hydromorphone through the G protein-over β-arrestin-mediated pathway: cAMP, CellKey™, and receptor internalization analyses.
Journal of pharmacological sciences (2019-07-20)
Sei Manabe, Kanako Miyano, Yuriko Fujii, Kaori Ohshima, Yuki Yoshida, Miki Nonaka, Miaki Uzu, Yoshikazu Matsuoka, Tetsufumi Sato, Yasuhito Uezono, Hiroshi Morimatsu
ABSTRACT
Morphine, fentanyl, and oxycodone are widely used as analgesics, and recently hydromorphone has been approved in Japan. Although all of these are selective for μ-opioid receptors (MORs) and have similar structures, their analgesic potencies and adverse effects (AEs) are diverse. Recent molecular analyses of MOR signaling revealed that the G protein-mediated signaling pathway causes analgesic effects and the β-arrestin-mediated signaling pathway is responsible for AEs. We used several cell-based analyses that selectively measure cellular responses activated by either G protein- or β-arrestin-mediated pathways. GloSensor™ cAMP, CellKey™, and receptor internalization assays were performed with four different types of cells stably expressing differentially labelled MOR. EC50 values measured by cAMP and CellKey™ assays had potencies in the order fentanyl ≤ hydromorphone < morphine ≤ oxycodone, all also exhibiting full agonist responses. However, in the internalization assay, only fentanyl elicited a full agonist response. Hydromorphone had the strongest potency next to fentanyl; however, contribution of the β-arrestin-mediated pathway was small, suggesting that its effect could be biased toward the G protein-mediated pathway. Based on these properties, hydromorphone could be chosen as an effective analgesic.