MilliporeSigma
  • Alterations in phospholipid profiles of erythrocytes deep-frozen without cryoprotectants.
Alterations in phospholipid profiles of erythrocytes deep-frozen without cryoprotectants.
Drug testing and analysis (2019-04-06)
Yoeseph Cho, Ji-Hye Woo, Oh-Seung Kwon, Sang Sun Yoon, Junghyun Son
ABSTRACT
The erythrocyte membrane is composed of a phospholipid bilayer, which is known to undergo physicochemical changes during storage at low temperatures. This study was conducted to identify marker phospholipids that indicate alteration during deep-frozen storage and to determine the amount of marker phospholipids. Our research suggested a method to detect phospholipids by profiling analysis of thermally injured red blood cells (RBCs) without protecting agents. Human blood was stored at -80°C for 72 days. The RBC membrane phospholipids were extracted through a modified Bligh and Dyer method. Six selected phospholipids were analyzed and quantified using liquid chromatography-tandem mass spectrometry, and an in vitro model system was developed. The intracellular level of N-nervonoyl-D-erythro-sphingosylphosphorylcholine significantly increased in the thermally injured RBCs, and multiple biomarker candidates were evaluated by profiling analysis and mass spectrometry technology for targeted metabolomics.