MilliporeSigma
  • Telbivudine can safely reduce mother-to-child transmission in chronic hepatitis B women after 12 weeks of gestation.
Telbivudine can safely reduce mother-to-child transmission in chronic hepatitis B women after 12 weeks of gestation.
BMC infectious diseases (2019-07-14)
Li-Fen Han, Jian-Ming Zheng, Li-Qing Zheng, Hai-Bing Gao, Li-Xia Chen, Qing-Ling Xu, Yi-Hong Chai, Xin Zhang, Chen Pan, Lv-Feng Yao
ABSTRACT
To evaluate the efficacy and safety of telbivudine in chronic hepatitis B women during the second and third trimesters of pregnancy. The week 12-34 of pregnant women were screened in this prospective non-intervention study, with HBV DNA > 106 IU/mL and alanine aminotransferase > 50 IU/L. The patients were received telbivudine treatment as a treatment group or without antiviral treatment as a control group. All infants were received recombinant hepatitis B vaccine 10 μg within 12 h of birth, at week 4 and week 24, immunoglobulin G within 12 h of birth and were detected HBV markers at the range from 7 to 12 months after delivery. A total of 241 patients were finally enrolled, 139 patients in telbivudine group and 102 patients in control group. HBsAg negative rate of infants was 99.3% (135/136) in telbivudine group and was 91.9% (91/99) in control group after 7 months (P = 0.005), respectively. The incidence of undetectable HBV DNA levels (47.5%) was significantly lower in telbivudine-treated mothers than that in the controls (0%), and 75.5% patients alanine aminotransferase returned to normal in telbivudine group, and 51% in control group at delivery (P < 0.001), respectively. Telbivudine can safely reduce mother-to-child transmission in chronic hepatitis B women after 12 weeks of gestation.