MilliporeSigma
  • Influence of a Pre-organized N-Donor Group on the Coordination of Trivalent Actinides and Lanthanides by an Aminopolycarboxylate Complexant.
Influence of a Pre-organized N-Donor Group on the Coordination of Trivalent Actinides and Lanthanides by an Aminopolycarboxylate Complexant.
Chemistry (Weinheim an der Bergstrasse, Germany) (2018-11-18)
Colt R Heathman, Travis S Grimes, Santa Jansone-Popova, Santanu Roy, Vyacheslav S Bryantsev, Peter R Zalupski
ABSTRACT
The thermodynamic influence of a pre-organized N-donor group on the coordination of trivalent actinides and lanthanides by an aqueous aminopolycarboxylate complexant has been investigated. The synthesized reagent, N-2-methylpicolinate-ethylenediamine-N,N',N'-triacetic acid (EDTA-Mpic), resembles ethylenediamine-N,N,N',N'-tetraacetic acid (EDTA) with a single acetate pendant arm replaced by a 6-carboxypyridin-2-ylmethyl group. The rigid N-donor picolinate functionality has a profound impact on ligand protonation and trivalent f element complexation equilibria, as demonstrated by potentiometric, spectroscopic, and liquid/liquid metal-partitioning studies as well as by molecular dynamics calculations. Relative to diethylenetriamine-N,N,N',N'',N''-pentaacetic acid (DTPA), the ability to preferentially bind trivalent actinides over trivalent lanthanides was moderately lowered due to the presence of the N-(6-carboxypyridin-2-ylmethyl) substituent. The structural modification substantially amplifies the total ligand acidity of EDTA-Mpic. As a result the complexant sustains the metal complexation and efficient An3+ /Ln3+ differentiation in aqueous mixtures of unprecedented acidity for this class of reagents.