MilliporeSigma
  • An integrated microfluidic system with field-effect-transistor sensor arrays for detecting multiple cardiovascular biomarkers from clinical samples.
An integrated microfluidic system with field-effect-transistor sensor arrays for detecting multiple cardiovascular biomarkers from clinical samples.
Biosensors & bioelectronics (2019-02-01)
Anirban Sinha, Tse-Yu Tai, Kuang-Hsien Li, Priya Gopinathan, Yi-Da Chung, Indu Sarangadharan, Hsi-Pin Ma, Po-Chiun Huang, Shu-Chu Shiesh, Yu-Lin Wang, Gwo-Bin Lee
ABSTRACT
Certain blood-borne biomarkers offer a potent methodology for understanding the risk of cardiovascular diseases (CVDs) with clinicians generally advocating the use of multiple biomarkers for proper risk assessment of CVDs. Herein four such CVDs biomarkers- C-reactive protein (CRP), N-terminal pro b-type natriuretic peptide (NT-proBNP), cardiac troponin I (cTnI), and fibrinogen- were rapidly (5 min) analyzed from clinical samples (~ 4 µL) on an integrated microfluidic platform equipped with 1) immobilized highly specific aptamer probes and 2) field-effect transistor (FET)-based sensor arrays. The calibration curve from the FET sensor arrays showed good agreement in the physiological concentration ranges for CRP (0.1-50 mg/L), NT-proBNP (50-10,000 pg/mL), cTnI (1-10,000 pg/mL), and fibrinogen (0.1-5 mg/mL). The developed prototype of this fully automated portable device requires minimal reagent and sample inputs and consequently shows great promise for next-generation point-of-care devices assaying multiple CVDs biomarkers in clinical samples.