MilliporeSigma
  • Novel one step transformation method for Escherichia coli and Staphylococcus aureus using arginine-glucose functionalized hydroxyapatite nanoparticles.
Novel one step transformation method for Escherichia coli and Staphylococcus aureus using arginine-glucose functionalized hydroxyapatite nanoparticles.
Materials science & engineering. C, Materials for biological applications (2019-01-05)
Ketaki Deshmukh, Sutapa Roy Ramanan, Meenal Kowshik
ABSTRACT
Bacterial gene transformation is one of the important techniques in molecular biology which has significant applications in gene cloning technology. In this study, we have developed arginine-glucose functionalized hydroxyapatite nanoparticle (R-G-HAp NPs) mediated novel one step transformation method, effective for both Gram-positive and Gram-negative bacteria. R-G-HAp NPs served as carriers to deliver pDNA into Escherichia coli and Staphylococcus aureus at room temperature, without the need for preparation of competent cells. High transformation efficiency was achieved in Gram-positive, S. aureus (107 cfu/μg of pDNA) as well as Gram-negative, E. coli (109 cfu/μg of pDNA). This demonstrates the efficacy of R-G-HAp NPs as a nano-vehicle to achieve high plasmid transformation efficiency, even in Gram-positive bacteria which is usually a challenge, exhibiting their potential as promising synthetic non-viral vectors for efficient bacterial gene transformation.