MilliporeSigma
  • Adenovirus oncoprotein E4orf6 triggers Cullin5 neddylation to activate the CLR5 E3 ligase for p53 degradation.
Adenovirus oncoprotein E4orf6 triggers Cullin5 neddylation to activate the CLR5 E3 ligase for p53 degradation.
Biochemical and biophysical research communications (2019-07-16)
Haoran Guo, Siyu Shen, Yan Li, Ran Bi, Nannan Zhang, Wenwen Zheng, Yuyou Deng, Ying Yang, Xiao-Fang Yu, Chunxi Wang, Wei Wei
ABSTRACT
The human adenovirus oncoprotein E4orf6 hijacks intracellular Cullin 5-based E3 ubiquitin ligases (CRL5s) to induce the degradation of host proteins, including p53, that impede efficient viral replication. The complex also relies on another viral protein, E1B55K, to recruit substrates for ubiquitination. However, the determinants of adenoviral E4orf6-CRL5 E3 ligase-mediated p53 degradation in the scaffolding protein Cullin5 remain rarely investigated. Here, we demonstrated that the viral protein E4orf6 triggered relocalization of the Cullin5 protein from the cytoplasm to the nucleus and induced activation of the CRL5 E3 ligase via facilitating neddylation. The expression of the deneddylase SENP8/Den1 was significantly downregulated by E4orf6. We then identified SENP8 as a natural restriction factor for E4orf6-induced p53 degradation. Furthermore, our results indicated that the NEDD8-conjugating E2 enzyme UBE2M was essential for E4orf6-mediated p53 degradation and that its dominant negative mutant UBE2M C111S dramatically blocked E4orf6 functions. The Nedd8-activating enzyme inhibitor MLN4924 decreased E4orf6-induced neddylation of the cullin5 protein and subsequently suppressed p53 degradation. Collectively, our findings illuminate the strategy by which this viral oncoprotein specifically utilizes the neddylation pathway to activate host CRL E3 ligases to degrade host restriction factors. Disrupting this post-translational modification is an attractive pharmacological intervention against human adenoviruses.