MilliporeSigma
  • Enhanced AMPA receptor-mediated excitatory transmission in the rodent rostromedial tegmental nucleus following lesion of the nigrostriatal pathway.
Enhanced AMPA receptor-mediated excitatory transmission in the rodent rostromedial tegmental nucleus following lesion of the nigrostriatal pathway.
Neurochemistry international (2018-11-19)
Yongli Chang, Chengxue Du, Lingna Han, Shuxuan Lv, Jin Zhang, Guanyun Bian, Guoyi Tang, Yiwei Liu, Tao Chen, Jian Liu
ABSTRACT
The GABAergic rostromedial tegmental nucleus (RMTg) has reciprocal connections with the dopaminergic ventral tegmental area and substantia nigra pars compacta (SNc), and is involved in inhibitory control of monoaminergic nuclei. At present, it is not clear whether unilateral 6-hydroxydopamine lesions of the SNc in rats affect AMPA receptor-mediated excitatory transmission in the RMTg. Here we found that lesions of the SNc in rats increased the firing rate of GABAergic neurons and the level of glutamate in the RMTg compared to sham-operated rats. Intra-RMTg injection of AMPA receptor agonist (S)-AMPA increased the firing rate of the GABAergic neurons in both sham-operated and the lesioned rats, while AMPA receptor antagonist NBQX decreased the firing rate of the neurons. Further, intra-RMTg injection of (S)-AMPA decreased the levels of dopamine and serotonin in the medial prefrontal cortex (mPFC) in the two groups of rats; conversely, NBQX increased the levels of dopamine and serotonin. Compared to sham-operated rats, the duration of (S)-AMPA and NBQX action on the firing rate of GABAergic neurons in the RMTg and release of doapmine and serotonin in the mPFC was prolonged in the lesioned rats. In addition, lesions of the SNc in rats increased protein expression of t-GluR1 and p-GluR1-S831 subunits compared to sham-operated rats. Therefore, these changes in the lesioned rats are associated with increased release of glutamate and up-regulated expression of GluR1 subunit-containing AMPA receptors in the RMTg, which suggest that degeneration of the nigrostriatal pathway enhances AMPA receptor-mediated excitatory transmission in the RMTg.