MilliporeSigma
  • Influence of Oligonucleotide Grafting Density on Surface-Mediated DNA Transport and Hybridization.
Influence of Oligonucleotide Grafting Density on Surface-Mediated DNA Transport and Hybridization.
ACS nano (2019-06-28)
Jeremiah C Traeger, Zachary Lamberty, Daniel K Schwartz
ABSTRACT
Adsorption of soluble DNA to surfaces decorated with complementary DNA plays an important role in many bionanotechnology applications, and previous studies have reported complex dependencies of the surface density of immobilized DNA on hybridization. While these effects have been speculatively ascribed to steric or electrostatic effects, the influence of surface-mediated molecular transport (i.e., intermittent "hopping diffusion") has not been fully appreciated. Here, single-molecule tracking and Förster resonance energy transfer (FRET) were employed to characterize the mobility and the hybridization efficiency of adsorbed ssDNA oligonucleotides ("target") at solid-liquid interfaces exhibiting surface-immobilized ssDNA ("probe") over a wide range of surface grafting densities. Two distinct regimes were observed, with qualitatively different transport and hybridization behaviors. At dilute grafting density, only 1-3% of target molecules were observed to associate with probes (i.e., to hybridize). Adsorbing target molecules often searched unsuccessfully and "flew", via desorption-mediated diffusion, to secondary locations before hybridizing. In contrast, at high probe grafting density, approximately 20% of target DNA hybridized to immobilized probes, and almost always in the vicinity of initial adsorption. Moreover, following a dehybridization event, target molecules rehybridized at high probe density, but rehybridization was infrequent in the dilute density regime. Interestingly, the intermittent interfacial transport of mobile target molecules was suppressed by the presence of immobilized probe DNA, presumably due to an increased probability of readsorption following each "hop". Together, these findings suggested that many salient effects of grafting density on surface-mediated DNA hybridization can be directly related to the mechanisms of surface-mediated intermittent diffusion.