MilliporeSigma
  • Molecular characterization and biological functioning of interleukin-8 in Siberian sturgeon (Acipenser baeri).
Molecular characterization and biological functioning of interleukin-8 in Siberian sturgeon (Acipenser baeri).
Fish & shellfish immunology (2019-04-13)
Xiaowen Wang, Guoqing Ma, Rong Zhang, Lili Liu, Jianya Zhu, Hua Zhu
ABSTRACT
Interleukin-8, otherwise known as CXCL8, is a CXC chemokine that plays a pivotal regulatory role in immune and inflammation responses of animals. Here, we identified an interleukin-8 homologue from Siberian sturgeon (Acipenser baeri), named AbIL-8, which belongs to the lineage 1 group of teleost fish IL-8s. The cDNA of Abil-8 is 1130 bp in length, containing a 5'- untranslated region (UTR) of 50 bp, a 3'- UTR of 783 bp, and an open reading frame (ORF) of 297 bp that encodes a protein consisting of 98 amino acids. The deduced AbIL-8 contained five cysteines, four of which are highly conserved, and an ELR motif typical of known mammalian CXC chemokines was also found preceding the CXC motif. Our phylogenetic analysis showed that AbIL-8 clustered with the CXCL8_L1 sequences from other teleosts, being clearly distinct from those of either birds or mammals. Abil-8 mRNA was constitutively expressed in all tested tissues and significantly up-regulated in the liver and spleen tissues by the bacteria Aernomas hydrophila. The in vitro experiment using primary spleen cells stimulated with heat-killed Aernomas hydrophila or lipopolysaccharide (LPS) revealed a similar expression pattern to that found in vivo, whereas stimulation on spleen cells with β-glucan or polyI:C elicited negligible changes in levels of Abil-8 mRNA. Purified recombinant AbIL-8 not only exhibited chemotactic activity for lymphocytes and monocytes in peripheral blood leukocytes (PBLs) and, to a lesser extent, spleen cells, but also stimulated the proliferation of spleen cells at 10 ng/mLor above. Furthermore, intraperitoneal injection of rAbIL-8 also up-regulated the expression of immuno-related genes (IL-6, IgM and MHCIIβ) at 24 h. Collectively, these results enhance our understanding of how IL-8 functions in the regulation of the immune responses in sturgeon.