MilliporeSigma
  • Spirodela polyrhiza and its Chemical Constituent Vitexin Exert Anti-Allergic Effect via ORAI1 Channel Inhibition.
Spirodela polyrhiza and its Chemical Constituent Vitexin Exert Anti-Allergic Effect via ORAI1 Channel Inhibition.
The American journal of Chinese medicine (2018-08-29)
Hyun Jong Kim, Yu Ran Nam, Eun-Jung Kim, Joo Hyun Nam, Woo Kyung Kim
ABSTRACT
Intracellular calcium signaling cascades are integral to early and late allergic responses involving mast cell degranulation and type 2 helper T cell activation, respectively. Both the responses are accompanied by the movement of calcium through the calcium release-activated calcium (CRAC) channel, encoded by the ORAI1 gene. Spirodela polyrhiza (L.) Schleid (SP) has anti-inflammatory and anti-allergic effects, but its effect on calcium signaling has not been reported. This study investigated whether a 30% ethanolic SP extract (SPEtOH) and its constituents can reduce CRAC currents ([Formula: see text]), and thus inhibit mast cell degranulation and T cell activation. In Jurkat T lymphocytes, we found that 3[Formula: see text]mg/mL SPEtOH inhibited the [Formula: see text] by [Formula: see text]%, whereas one of its constituents vitexin (100[Formula: see text][Formula: see text]M) inhibited the [Formula: see text] by [Formula: see text]%. Furthermore, in the RBL-2H3 mast cell, the [Formula: see text] was inhibited by 3[Formula: see text]mg/mL SPEtOH ([Formula: see text]%) and 100[Formula: see text][Formula: see text]M vitexin ([Formula: see text]%). Investigation of human primary T cell proliferation induced by co-stimulation with antibodies to cluster of differentiation 3 and 28, and of RBL-2H3 mast cell degranulation following IgE-antigen complex stimulation revealed that 100[Formula: see text][Formula: see text]M vitexin inhibited both T-cell proliferation (by [Formula: see text]%) and mast cell degranulation (by [Formula: see text]%). These effects were concentration-dependent, and no cytotoxicity was observed. Our findings suggest that vitexin is a promising candidate compound for the development of therapeutic agents to prevent and treat allergic diseases.