MilliporeSigma
  • PARPi Triggers the STING-Dependent Immune Response and Enhances the Therapeutic Efficacy of Immune Checkpoint Blockade Independent of BRCAness.
PARPi Triggers the STING-Dependent Immune Response and Enhances the Therapeutic Efficacy of Immune Checkpoint Blockade Independent of BRCAness.
Cancer research (2018-11-30)
Jianfeng Shen, Wei Zhao, Zhenlin Ju, Lulu Wang, Yang Peng, Marilyne Labrie, Timothy A Yap, Gordon B Mills, Guang Peng
ABSTRACT
PARP inhibitors (PARPi) have shown remarkable therapeutic efficacy against BRCA1/2-mutant cancers through a synthetic lethal interaction. PARPi exert their therapeutic effects mainly through the blockade of ssDNA damage repair, which leads to the accumulation of toxic DNA double-strand breaks specifically in cancer cells with DNA repair deficiency (BCRAness), including those harboring BRCA1/2 mutations. Here we show that PARPi-mediated modulation of the immune response contributes to their therapeutic effects independently of BRCA1/2 mutations. PARPi promoted accumulation of cytosolic DNA fragments because of unresolved DNA lesions, which in turn activated the DNA-sensing cGAS-STING pathway and stimulated production of type I IFNs to induce antitumor immunity independent of BRCAness. These effects of PARPi were further enhanced by immune checkpoint blockade. Overall, these results provide a mechanistic rationale for using PARPi as immunomodulatory agents to harness the therapeutic efficacy of immune checkpoint blockade. SIGNIFICANCE: This work uncovers the mechanism behind the clinical efficacy of PARPi in patients with both BRCA-wild-type and BRCA-mutant tumors and provides a rationale for combining PARPi with immunotherapy in patients with cancer.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Tubulin γ, C-Terminal antibody produced in rabbit, affinity isolated antibody