MilliporeSigma
  • Human UTP14a promotes colorectal cancer progression by forming a positive regulation loop with c-Myc.
Human UTP14a promotes colorectal cancer progression by forming a positive regulation loop with c-Myc.
Cancer letters (2018-10-22)
Jingyi Zhang, Pengwei Ren, Da Xu, Xiaofeng Liu, Zhenzhen Liu, Chunfeng Zhang, Yuan Li, Lijun Wang, Xiaojuan Du, Baocai Xing
ABSTRACT
Nucleolar protein hUTP14a is required for 18S rRNA processing and promotes p53 degradation. Here, we report that hUTP14a stabilizes c-Myc in colorectal cancer (CRC) progression. Firstly, nucleolar hUTP14a is upregulated in human CRC tissues. Mass spectrometry analysis identified c-Myc and its deubiquitinase ubiquitin-specific protease 36 (USP36) in the hUTP14a-specific complex. Importantly, hUTP14a interacts with c-Myc and protects c-Myc from ubiquitination and degradation in a USP36-dependent way. We further demonstrate that hUTP14a forms a complex with USP36/Fbw7γ to inhibit Fbw7γ-mediated c-Myc degradation. Ectopic expression of Flag-hUTP14a enriches c-Myc in the nucleolus, indicating hUTP14a stabilizes c-Myc in the nucleolus. Interestingly, c-Myc activates transcription of hUTP14a. Knockdown of hUTP14a by short hairpin RNA inhibits tumor growth and decreases c-Myc levels in mouse xenografts. Significantly, nucleolar hUTP14a and c-Myc are co-upregulated in human CRC tissues, and this co-upregulation indicates poor prognosis of CRC patients. Thus, disruption of hUTP14a-c-Myc regulation may provide a potential therapeutic strategy for a subset of CRC patients.