MilliporeSigma
  • Antifungal Activity against Botrytis cinerea of 2,6-Dimethoxy-4-(phenylimino)cyclohexa-2,5-dienone Derivatives.
Antifungal Activity against Botrytis cinerea of 2,6-Dimethoxy-4-(phenylimino)cyclohexa-2,5-dienone Derivatives.
Molecules (Basel, Switzerland) (2019-02-20)
Paulo Castro, Leonora Mendoza, Claudio Vásquez, Paz Cornejo Pereira, Freddy Navarro, Karin Lizama, Rocío Santander, Milena Cotoras
ABSTRACT
In this work the enzyme laccase from Trametes versicolor was used to synthetize 2,6-dimethoxy-4-(phenylimino)cyclohexa-2,5-dienone derivatives. Ten products with different substitutions in the aromatic ring were synthetized and characterized using ¹H- and 13C-NMR and mass spectrometry. The 3,5-dichlorinated compound showed highest antifungal activity against the phytopathogen Botrytis cinerea, while the p-methoxylated compound had the lowest activity; however, the antifungal activity of the products was higher than the activity of the substrates of the reactions. Finally, the results suggested that these compounds produced damage in the fungal cell wall.