MilliporeSigma
  • Effects of elfamycins on elongation factor Tu from Escherichia coli and Staphylococcus aureus.
Effects of elfamycins on elongation factor Tu from Escherichia coli and Staphylococcus aureus.
Antimicrobial agents and chemotherapy (1989-03-01)
C C Hall, J D Watkins, N H Georgopapadakou
ABSTRACT
Six kirromycin analogs (elfamycins) were compared on the basis of their inhibition of Escherichia coli poly(U)-directed poly(Phe) synthesis and stimulation of elongation factor Tu (EF-Tu)-associated GTPase activity. The elfamycins tested were kirromycin, aurodox, efrotomycin, phenelfamycin A, unphenelfamycin, and L-681,217. The last three lack the pyridone ring present in the other elfamycins. All the elfamycins inhibited poly(U)-dependent poly(Phe) synthesis and stimulated EF-Tu-associated GTPase activity, suggesting that the pyridone ring is not essential for activity. The six elfamycins were also examined in a poly(U)-directed, poly(Phe)-synthesizing system derived from Staphylococcus aureus and had 50% inhibitory concentrations of greater than or equal to 1 mM. When S. aureus ribosomes and E. coli elongation factors were combined in a hybrid poly(Phe)-synthesizing system, aurodox produced essentially complete inhibition of poly(Phe) synthesis with a 50% inhibitory concentration of 0.13 microM. This suggests that the observed high MICs of kirromycin and its congeners in S. aureus reflect a kirromycin-resistant EF-Tu rather than permeability constraints.