MilliporeSigma
  • Safety assessment of antioxidants and color fixatives for the Korean population using dietary intake monitoring.
Safety assessment of antioxidants and color fixatives for the Korean population using dietary intake monitoring.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2019-04-16)
Gunyoung Lee, Ho Soo Lim, Sang Soon Yun, Jae-Wook Shin, MeeKyung Kim
ABSTRACT
Food additives such as antioxidants and color fixatives are substances used in food intentionally for technical effect, such as decolorizing or intensifying the color of food. Based on the necessity of re-evaluating food additives for safety and to improve consumer perception, we conducted safety assessments for food additives according to the Risk Assessment Guidelines of the Korean Ministry of Food and Drug Safety. These safety assessments evaluated new risk information based on toxicology data and estimates of dietary intake exposures to food additives in comparison with the acceptable daily intake (ADI). Estimated daily intakes (EDI) of food additives were calculated using food consumption data for the Korean population derived from the Korea National Health and Nutrition Examination Survey and monitoring data based on the analysis of food additives in food products. Unlike contaminants, antioxidants and color fixatives are purposely added as food additives, and they are largely consumed in processed foods. Therefore, EDI was compared with ADI to investigate the likelihood of potentially hazardous effects in humans. The risk likelihoods of food additives, evaluated by comparing the EDI with the ADI, were less than 2% in the total population. Thus, exposure levels to antioxidants and color fixatives do not exceed the ADI. Based on the safety assessments conducted in this study, we estimate exposure to food additives to be within safe limits for all population groups.