MilliporeSigma
  • Cutaneous T-Cell Lymphoma: Optimizing Care in Patients Receiving Anti-CCR4 Monoclonal Antibody Mogamulizumab.
Cutaneous T-Cell Lymphoma: Optimizing Care in Patients Receiving Anti-CCR4 Monoclonal Antibody Mogamulizumab.
Clinical journal of oncology nursing (2019-07-20)
Marianne Tawa, Erin Kopp, Sue McCann, Wendy Cantrell
ABSTRACT
Cutaneous T-cell lymphoma (CTCL), including subtypes mycosis fungoides (MF) and Sézary syndrome (SS), represents a rare group of non-Hodgkin lymphomas. Mogamulizumab is a first-in-class monoclonal antibody that selectively binds to C-C chemokine receptor 4, which is overexpressed on the surface of tumor cells in T-cell malignancies, including MF/SS-type CTCL. This review identifies common diagnostic features of MF/SS, the efficacy and side effect profile of mogamulizumab, and practical management strategies for optimizing the nursing care of patients with MF/SS-type CTCL. Case studies are used to describe the role of mogamulizumab in CTCL and to review practical considerations when administering mogamulizumab to patients. Mogamulizumab is an effective treatment for adult patients with relapsed or refractory MF/SS-type CTCL who have received at least one prior systemic therapy. Infusion reactions and drug eruptions require prompt diagnosis and treatment.