MilliporeSigma
  • Arsenic Speciation on Silver Nanofilms by Surface-Enhanced Raman Spectroscopy.
Arsenic Speciation on Silver Nanofilms by Surface-Enhanced Raman Spectroscopy.
Analytical chemistry (2019-06-15)
Mingwei Yang, Valery Liamtsau, Changjun Fan, Kelli L Sylvers, Anthony J McGoron, Guangliang Liu, Fengfu Fu, Yong Cai
ABSTRACT
Surface-enhanced Raman spectroscopy (SERS), as a nondestructive and fast detection technique, is a promising alternative approach for arsenic detection, particularly for in situ applications. SERS-based speciation analysis according to the fingerprint SERS signals of different arsenicals has the potential to provide a superior technique in species preservation over the conventional chromatographic separation methods, albeit with some difficulties due to the similarity in SERS patterns. In this study, we explored a novel SERS method for arsenic speciation by using the separation potential of the coffee ring effect on negatively charged silver nanofilms (AgNFs). Four arsenic species, including arsenite (AsIII), arsenate (AsV), monomethylarsonic acid (MMAV), and dimethylarsinic acid (DMAV), were measured for fingerprint SERS signals in solution and on the films. Significant enhancement of SERS signals on the dried coffee ring stains by the AgNFs were observed except for AsIII, and more importantly, arsenicals migrated varying distances during coffee ring development, promoting better speciation. Sodium dodecyl sulfate was then introduced into the droplet to reduce the droplet surface tension, facilitating the migration of solution into the peripheral region. Under the combined interactions of arsenicals with the AgNFs, solvent, and surfactant, enhanced separation between arsenicals was observed as a result of the formation of two concentric rings. Combining the SERS fingerprint signals and physical separation of arsenicals on the surface, arsenic speciation was achieved using the AgNFs substrate-based SERS technology, demonstrating the potential of the coffee ring effect for rapid separation and analysis of small molecules by SERS.