MilliporeSigma
  • Serum Metabolic Profiling Identified a Distinct Metabolic Signature in Bladder Cancer Smokers: A Key Metabolic Enzyme Associated with Patient Survival.
Serum Metabolic Profiling Identified a Distinct Metabolic Signature in Bladder Cancer Smokers: A Key Metabolic Enzyme Associated with Patient Survival.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2019-01-16)
Chandra Sekhar Amara, Chandrashekar R Ambati, Venkatrao Vantaku, Danthasinghe Waduge Badrajee Piyarathna, Sri Ramya Donepudi, Shiva Shankar Ravi, James M Arnold, Vasanta Putluri, Gurkamal Chatta, Khurshid A Guru, Hoda Badr, Martha K Terris, Roni J Bollag, Arun Sreekumar, Andrea B Apolo, Nagireddy Putluri
ABSTRACT
The current system to predict the outcome of smokers with bladder cancer is insufficient due to complex genomic and transcriptomic heterogeneities. This study aims to identify serum metabolite-associated genes related to survival in this population. We performed LC/MS-based targeted metabolomic analysis for >300 metabolites in serum obtained from two independent cohorts of bladder cancer never smokers, smokers, healthy smokers, and healthy never smokers. A subset of differential metabolites was validated using Biocrates absoluteIDQ p180 Kit. Genes associated with differential metabolites were integrated with a publicly available cohort of The Cancer Genome Atlas (TCGA) to obtain an intersecting signature specific for bladder cancer smokers. Forty metabolites (FDR < 0.25) were identified to be differential between bladder cancer never smokers and smokers. Increased abundance of amino acids (tyrosine, phenylalanine, proline, serine, valine, isoleucine, glycine, and asparagine) and taurine were observed in bladder cancer smokers. Integration of differential metabolomic gene signature and transcriptomics data from TCGA cohort revealed an intersection of 17 genes that showed significant correlation with patient survival in bladder cancer smokers. Importantly, catechol-O-methyltransferase, iodotyrosine deiodinase, and tubulin tyrosine ligase showed a significant association with patient survival in publicly available bladder cancer smoker datasets and did not have any clinical association in never smokers. Serum metabolic profiling of bladder cancer smokers revealed dysregulated amino acid metabolism. It provides a distinct gene signature that shows a prognostic value in predicting bladder cancer smoker survival. Serum metabolic signature-derived genes act as a predictive tool for studying the bladder cancer progression in smokers.