MilliporeSigma
  • Ion-chromatographic determination of carbocisteine in pharmaceuticals based on non-suppressed conductimetric detection
Ion-chromatographic determination of carbocisteine in pharmaceuticals based on non-suppressed conductimetric detection
Journal of Chromatography A (2004)
Megoulas NC and Koupparis MA