MilliporeSigma
  • Determination of nimesulide in pharmaceutical dosage forms by second order derivative UV spectrophotometry
Determination of nimesulide in pharmaceutical dosage forms by second order derivative UV spectrophotometry
Journal of Pharmaceutical and Biomedical Analysis (2000)
Altinoz S and Dursun OO