MilliporeSigma
  • An isocratic HPLC method for the determination of sorbitol and glycerol in pharmaceutical formulations
An isocratic HPLC method for the determination of sorbitol and glycerol in pharmaceutical formulations
Journal of Chromatographic Science (2012)
Simonzadeh N and Ronsen B