MilliporeSigma
  • Spectrophotometric determination of omeprazole, lansoprazole and pantoprazole in pharmaceutical formulations
Spectrophotometric determination of omeprazole, lansoprazole and pantoprazole in pharmaceutical formulations
Journal of Pharmaceutical and Biomedical Analysis (2002)
Wahbi AAM, et al.