MilliporeSigma
  • Purification of dipeptidyl aminopeptidase II (dipeptidyl arylamidase II) of the anterior pituitary gland. Peptidase and dipeptide esterase activities.
Purification of dipeptidyl aminopeptidase II (dipeptidyl arylamidase II) of the anterior pituitary gland. Peptidase and dipeptide esterase activities.
The Journal of biological chemistry (1968-08-10)
J K McDonald, F H Leibach, R E Grindeland, S Ellis