MilliporeSigma
  • The effects of cell death-inducing DNA fragmentation factor-alpha-like effector C (CIDEC) on milk lipid synthesis in mammary glands of dairy cows
The effects of cell death-inducing DNA fragmentation factor-alpha-like effector C (CIDEC) on milk lipid synthesis in mammary glands of dairy cows
Journal of Dairy Science (2017)
Yang Y, et al.