MilliporeSigma
  • Epitopes resistance to the simulated gastrointestinal digestion of beta-lactoglobulin submitted to two-step enzymatic modification
Epitopes resistance to the simulated gastrointestinal digestion of beta-lactoglobulin submitted to two-step enzymatic modification
Food Research International (2015)
Villas-Boas M, et al.