MilliporeSigma
  • Efficient site-specific processing of fusion proteins by tobacco vein mottling virus protease in vivo and in vitro
Efficient site-specific processing of fusion proteins by tobacco vein mottling virus protease in vivo and in vitro
Protein Expression and Purification (2004)
Nallamsetty S, et al.