MilliporeSigma
  • Connexin43 forms supramolecular complexes through non-overlapping binding sites for drebrin, tubulin, and ZO-1
Connexin43 forms supramolecular complexes through non-overlapping binding sites for drebrin, tubulin, and ZO-1
Testing (2016)
Ambrosi C, et al.