MilliporeSigma
  • Structural basis for substrate specificity of Escherichia coli purine nucleoside phosphorylase
Structural basis for substrate specificity of Escherichia coli purine nucleoside phosphorylase
The Journal of biological chemistry (2003)
Bennett EM, et al.