MilliporeSigma
  • Measurement of nonribosomal peptide Synthetase Adenylation domain activity using a continuous hydroxylamine release assay
Measurement of nonribosomal peptide Synthetase Adenylation domain activity using a continuous hydroxylamine release assay
Methods in Molecular Biology (2016)
Duckworth BP, et al.