MilliporeSigma
  • A continuous kinetic assay for adenylation enzyme activity and inhibition
A continuous kinetic assay for adenylation enzyme activity and inhibition
Analytical Biochemistry (2010)
Wilson DJ and Aldrich CC