MilliporeSigma
  • Tolerance mechanisms in mercury-exposed Chromolaena odorata (lf) RM King et H. Robinson, a potential phytoremediator
Tolerance mechanisms in mercury-exposed Chromolaena odorata (lf) RM King et H. Robinson, a potential phytoremediator
Journal of Degraded and Mining Lands Management (2013)
Alcantara HJP, et al.