MilliporeSigma
  • A highly organized three-dimensional alginate scaffold for cartilage tissue engineering prepared by microfluidic technology
A highly organized three-dimensional alginate scaffold for cartilage tissue engineering prepared by microfluidic technology
Biomaterials (2011)
Wang CC, et al.