MilliporeSigma
  • Hormonal control of pyruvate kinase activity and of gluconeogenesis in isolated hepatocytes
Hormonal control of pyruvate kinase activity and of gluconeogenesis in isolated hepatocytes
Proceedings of the National Academy of Sciences of the USA (1976)
Feliu J E, et al.