MilliporeSigma
  • The proto-oncoprotein SYT (SS18) controls ATP release and regulates cyst formation by polarized MDCK cells
The proto-oncoprotein SYT (SS18) controls ATP release and regulates cyst formation by polarized MDCK cells
Experimental Cell Research (2008)
Chittezhath M, et al.