MilliporeSigma
  • Evaluation of the euglycemic effect of oral administration of S. rebaudiana B. cultivated in Mexico in normoglycemic and induced-diabetic rats
Evaluation of the euglycemic effect of oral administration of S. rebaudiana B. cultivated in Mexico in normoglycemic and induced-diabetic rats
International journal of environment, agriculture and biotechnology
Irma AG, et al.