MilliporeSigma
  • Determination of rebaudioside A and Stevioside in leaves of S. rebaudiana Bertoni grown in Mexico by a validated HPLC method
Determination of rebaudioside A and Stevioside in leaves of S. rebaudiana Bertoni grown in Mexico by a validated HPLC method
American Journal of Analytical Chemistry (2015)
Aranda-Gonzalez I, et al.