MilliporeSigma
  • Detection of Antibodies Against DNA Polymerase of Hepatitis B Virus in HBsAg-Positive Sera Using ELISA
Detection of Antibodies Against DNA Polymerase of Hepatitis B Virus in HBsAg-Positive Sera Using ELISA
The Korean Journal of Internal Medicine (1998)
Rui LX, et al.