MilliporeSigma
  • Analysis of alpha-lactalbumin, beta-lactoglobulin A and B in whey protein powder, colostrum, raw milk, and infant formula by CE and LC
Analysis of alpha-lactalbumin, beta-lactoglobulin A and B in whey protein powder, colostrum, raw milk, and infant formula by CE and LC
Dairy science & technology (2011)
Ding X, et al.